Learn More
HP Rotaflo™, Stopcock Spare Parts - Replacement Keys
Product Code. 12028743
Change view
Click to view available options
Plug Bore:
3 mm
6 mm
10 mm
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore Unit Size
12028743 10 mm Each
10719555 3 mm Pack of 2
12018743 6 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12028743 Supplier Quickfit Supplier No. HP10K
  • ONLINE EXCLUSIVE PRICE
    82.25 EUR
    68.11 EUR / Pack of 1
    Save 14.14 EUR
    17% Off
Estimated Shipment: 23-03-2027
Replacement keys for stopcock
  • Replacement keys for stopcock
Specifications
Type Stopcock Key
Plug Bore 10 mm
For Use With (Equipment) HP Rotaflo™ stopcock