Learn More
Sperian™ Honeywell™ Howard Leight™ Bilsom™ 303L Foam Earplugs
Product Code. 11827020
Change view
Click to view available options
Quantity:
200/Pk.
Unit Size:
200 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11827020 200/Pk. 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11827020 Supplier Sperian™ Supplier No. 1005073
  • ONLINE EXCLUSIVE PRICE
    74.32 EUR
    44.18 EUR / 200 pairs
    Save 30.14 EUR
    40% Off
Estimated Shipment: 16-12-2026
Ideal for workers who need to block out loud noise. Sperian™ Honeywell™ Howard Leight™ Bilsom™ 303L Foam Earplugs offer the best protection and comfort available in an earplug.
  • Soft polyurethane foam delivers superior long-wearing comfort and solid performance
  • Smooth, soil-resistant skin prevents dirt or other substances from accumulating on skin or entering ear canal
  • Europe's most-used polyurethane earplug
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow, White
Size Regular
Quantity 200/Pk.
Plug Shape Bullet
Product Type Ear Plugs