Learn More
Fisherbrand™ Hose Adapter Kit for Rotary Vane Vacuum Pumps
Product Code. 15576335 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15576335 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15576335
  • ONLINE EXCLUSIVE PRICE
    345.00 EUR
    321.27 EUR / Pack of 1
    Save 23.73 EUR
    6% Off
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
Use this hose adaptor kit with all Fisherbrand rotary van vacuum pumps. Fisherbrand™ Hose Adapter Kits for Rotary Vane Vacuum Pumps include stainless steel tubing adaptors and clamps for vacuum tight connections.
  • Vacuum hose adapter kit
  • For Fisherbrand rotary vane vacuum pumps
  • Stainless steel tubing adaptors and clamps for vacuum tight connections
  • 1.5m of thick walled rubber hose in Sizes DN10 and DN20
TRUSTED_SUSTAINABILITY
Specifications
Includes DN10 & DN20 hose, Stainless Steel adapters and clamps
Pump Type Hose Adapter Kit for Rotary Vane Vacuum Pumps
For Use With (Equipment) All Fisherbrand vacuum pumps