Learn More
Homatropine methylbromide, MedChemExpress
Product Code. 16494587
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Product Code. 16494587
Brand: Medchem Express HYB1388
MedChemExpress Homatropine methylbromide (Homatropine methobromide) is muscarinic AChR antagonist, inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 of 162.5 nM and 170.3 nM, respectively.
Homatropine methylbromide (Homatropine methobromide) is muscarinic AChR antagonist, inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 of 162.5 nM and 170.3 nM, respectively.
Specifications
Chemical Name or Material Homatropine methylbromide
CAS 80-49-9
Color White
Purity Grade Notes Research
Molecular Formula C17H24BrNO3
Quantity 50 mg
Synonym Homatropine methobromide
Solubility Information DMSO : ≥ 31 mg/mL (83.72 mM)
SMILES C[N+]1([C@@H]2C[C@@H](OC(C(O)C3=CC=CC=C3)=O)C[C@H]1CC2)C.[Br-]
Molecular Weight (g/mol) 370.28
Formula Weight 370.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation