Learn More
IKA™ H 38 Holding Rod
Product Code. 10043852 Shop All IKA Products
Change view
Click to view available options
Length (Metric) Rod:
184.5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Rod Unit Size
10043852 184.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10043852 Supplier IKA™ Supplier No. 0003547700
  • ONLINE EXCLUSIVE PRICE
    40.45 EUR
    39.00 EUR / Pack of 1
    Save 1.45 EUR
    3% Off
Estimated Shipment: 16-12-2026
Use this holding rod with support rod H 16 V and a temperature sensor or ETS-D5/6 electronic contact thermometer. IKA™ H 38 Holding Rod fastens a temperature sensor or ETS-D5/6 with a boss head H 44 to the support rod H 16 V.
  • For use with IKA electronic thermometers, support rods and temperature sensors
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) Rod 184.5 mm
Product Type Holding Rod
Size 10 mm dia.
For Use With (Application) Fastening ETS-D5 with H 44 to Support Rod
Quantity 1 Ea.