Learn More
Hoefer™ SG Gradient Maker: Outlet Fitting
Click to view available options
For Use With (Equipment):
SG 5, SG 15, SG 30, SG 50 or SG 100 Gradient Maker
Unit Size:
Each
Description
For use with SG Gradient Makers
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | SG 5, SG 15, SG 30, SG 50 or SG 100 Gradient Maker |
| Type | Luer Outlet Fitting |