Learn More
Hoefer™ SE 600 Chroma Vertical Electrophoresis System
Product Code. 11995418
Change view
Click to view available options
Voltage:
1000 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
11995418 1000 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11995418 Supplier Hoefer™ Supplier No. SE600X151.5
  • ONLINE EXCLUSIVE PRICE
    2025.00 EUR
    1912.04 EUR / Pack of 1
    Save 112.96 EUR
    5% Off
View all promotions
Estimated Shipment: 04-02-2027
Built-in heat exchanger maintains temperature between 1° and 45°C (33.8° and 113°F) for faster runs with superior quality results
  • SE600 Chroma has two built-in side handles and rounded contours that ease handling, lifting, introducing sample and disassembling
  • Run one gel or two gels simultaneously, four with optional divider plates
  • Produce straight lanes and sharp, well-defined bands—no “smiling”
  • Provide two mechanisms to dissipate heat
  • Gel sandwiches are completely immersed in the lower buffer so that heat is transferred evenly on both sides of the gel
  • A heat exchanger suspended vertically in the lower chamber cools or heats buffer evenly
Suitable for denaturing and native polyacrylamide, 2-D, isoelectric focusing and agarose gel electrophoresis
Certifications
UL3101-1, CSA C22.2 1010.1, CE Certified, EN61010-1
Specifications
Certifications/Compliance EN61010-1, UL61010-1, CSA C22.2 1010.1, CE
Humidity Range Up to 80%
Includes Lower Buffer chamber, upper Buffer chamber, safety lid with high voltage Leads, heat exchanger, Glass plates, clamp assemblies, cams, slotted gaskets for upper Buffer chamber, dual gel casting stand with leveling base, bubble level, laminated gaskets for
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Temperature (Metric) Storage 15°C to 30°C
Thickness (English) Spacer 0.05 in.
Thickness (Metric) Spacer 0.15 cm
Volume (English) Buffer 169.07 oz.
Volume (Metric) Buffer 5000 mL
Width (English) 12.59 in.
Width (English) Gel 7.08 in.
Width (Metric) 32 cm
Width (Metric) Gel 18 cm
Height (English) 11.41 in.
Height (English) Gel 6.29/3.14 in.
Height (Metric) 29 cm
Depth (English) 5.51 in.
Depth (Metric) 14 cm
Voltage 1000 V
Wattage 50 w
Amperage 500 mA