Learn More
Hoefer™ SE 400 Series Sturdier™ Air-Cooled Vertical Electrophoresis Systems
Product Code. 11925418
Change view
Click to view available options
Gel Dimensions:
18 x 16 cm
18 x 24 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Dimensions Unit Size
11925418 18 x 24 cm Each
10586935 18 x 16 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11925418 Supplier Hoefer™ Supplier No. SE410
  • ONLINE EXCLUSIVE PRICE
    1520.00 EUR
    1434.03 EUR / Pack of 1
    Save 85.97 EUR
    5% Off
Estimated Shipment: 18-02-2027
Includes
Upper Buffer chamber, safety lid with electrodes, lower Buffer chamber and casting stand on leveling base with high voltage Leads, Glass plates, clamp assemblies, cams, slotted gasket for UBC, bubble level, laminated gasket for casting stand, spacer-mate-alignment template, wonder wedge-plate separation tool, gel seal
Requires
Comb and spacers must be ordered separately for Model SE 410 Basic.
Economical units ideal for applications in which temperature control is not crucial
  • Units run one gel, or two with optional divider plate
  • Built-in casting stand seals sandwiches leak-free without tape
  • Rugged injection-molded construction for easy assembly and durability in high-use labs
  • Plates, clamps, spacers and combs are interchangeable with SE 600 series units
Certifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE Marked.
Specifications
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Gel Dimensions 18 x 24 cm
Humidity Range Up to 80%
Includes Upper Buffer chamber, safety lid with electrodes, lower Buffer chamber and casting stand on leveling base with high voltage Leads, Glass plates, clamp assemblies, cams, slotted gasket for UBC, bubble level, laminated gasket for casting stand, spacer-mate-alignment template, wonder wedge-plate separation tool, gel seal
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Temperature (Metric) Storage 15°C to 30°C
Volume (English) Buffer 23.66 oz.
Volume (Metric) Buffer 700 mL
Width (English) 9.44 in.
Width (English) Gel 7.08 in.
Width (Metric) 24 cm
Width (Metric) Gel 18 cm
Height (English) 14.17 in.
Height (English) Gel 9.44 in.
Height (Metric) 36 cm
Depth (English) 5.9 in.
Depth (Metric) 15 cm
Voltage 500 V
Wattage 20 w
Amperage 60 mA