Learn More
Hoefer™ HE 33 Mini Submarine Electrophoresis Unit
Product Code. 10646253
Change view
Click to view available options
Model:
Hoefer HE 33 Basic
Hoefer HE 33 Complete
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
10646253 Hoefer HE 33 Basic Each
10075904 Hoefer HE 33 Basic Each
10754807 Hoefer HE 33 Complete Each
11589533 Hoefer HE 33 Complete Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10646253 Supplier Hoefer™ Supplier No. HE33B
  • ONLINE EXCLUSIVE PRICE
    449.00 EUR
    423.90 EUR / Pack of 1
    Save 25.10 EUR
    5% Off
View all promotions
Estimated Shipment: 27-01-2027
Includes
Buffer chamber, lid with cables, UV-transparent gel running tray, gel casting tray, 4 foam gaskets, bubble level. Complete system (03-500-130) also includes 1.5mm thick 8-well comb and comb back.
Designed for very fast separations of DNA restriction fragments in agarose gels
  • The clear molded base acts as a heat-sink—simply fill the base with coolant and seal
  • Cooling the unit prior to a run allows higher voltages to be applied without generating band distortions or damaging the gel and unit
  • No taping required for casting
  • Electrical safety locks in lid
Specifications
Certifications/Compliance EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Type Mini Horizontal Agarose Electrophoresis Unit
Humidity Range Up to 80%
Includes Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level
Model Hoefer HE 33 Basic
Sample Size Up to 32
Temperature (English) Maximum 122°F
Temperature (Metric) Maximum 50°C
Volume (English) Buffer 8.4 oz.
Volume (Metric) Buffer 250 mL
Dimensions (L x W x H) 9.4 x 5.1 x 2.8 in. (24 x 13 x 7 cm)
Length (English) 9.4 in.
Length (English) Gel 3.9 in.
Length (Metric) 24 cm
Length (Metric) Gel 10 cm
Width (English) 5.1 in.
Width (English) Gel 2.7 in.
Width (Metric) 13 cm
Width (Metric) Gel 7 cm
Height (English) 2.8 in.
Height (Metric) 7 cm
Voltage 500 V
Wattage 15 w
Amperage 500 mA