Learn More
Hoefer™ GD 2000 Vacuum Gel Dryer Accessories: Rubber Overlay Sheet
Product Code. 11559633
Change view
Click to view available options
For Use With (Equipment):
GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11559633 GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11559633 Supplier Hoefer™ Supplier No. SE1143
  • ONLINE EXCLUSIVE PRICE
    118.00 EUR
    102.20 EUR / Pack of 1
    Save 15.80 EUR
    13% Off
Estimated Shipment: 05-01-2027
Dries acrylamide and agarose gels rapidly, evenly and safely.
  • Clear Silicone Rubber Overlay Sheet
Specifications
Color Clear
For Use With (Equipment) GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers
Type Rubber Overlay Sheet
Material Silicone