missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress HOE-S 785026 trihydrochloride

Product Code. 16476806
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476806 100 mg 100mg
16476628 10 mg 10mg
16419197 5 mg 5mg
16402727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476806 Supplier MedChemExpress Supplier No. HY15561B
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress HOE-S 785026 trihydrochloride is a blue fluorescent dyes, which can be used as a cell dye for DNA.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 100 mg
Shipping Condition Room temperature
Solubility H2O : 47.3 mg/mL (88.60 mM; Need ultrasonic and warming)
Molecular Weight (g/mol) 533.88 g/mol
Synonym meta-Hoechst trihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.