Learn More
hnRNP L Antibody (4D11), CoraFluor™ 1, Novus Biologicals™
Product Code. 30148071 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30148071 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30148071 Supplier Novus Biologicals Supplier No. NB1206106CL1
Mouse Monoclonal Antibody
hnRNP L Monoclonal antibody specifically detects hnRNP L in Amphibian,Bovine,Chicken,Human,Mouse,Primate,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Frozen),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Antigen hnRNP L
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 4D11
Conjugate CoraFluor 1
Formulation PBS
Gene Alias FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL
Host Species Mouse
Immunogen Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3191
Target Species Amphibian, Bovine, Chicken, Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG1