missing translation for 'onlineSavingsMsg'
Learn More

HMR 1556, MedChemExpress

Product Code. 16456787
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456787 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16456787 Supplier Medchem Express Supplier No. HY1063695mg
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress HMR 1556, a chromanol derivative, is a potent IKs blocker with IC50s of 10.5 nM and 34 nM in canine and guinea pig left ventricular myocytes, respectively.

HMR 1556, a chromanol derivative, is a potent IKs blocker with IC50s of 10.5 nM and 34 nM in canine and guinea pig left ventricular myocytes, respectively.

Specifications

Chemical Name or Material HMR 1556
CAS 223749-46-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H24F3NO5S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (243.05 mM; Need ultrasonic)
SMILES CS(N([C@H]1C2=CC(OCCCC(F)(F)F)=CC=C2OC(C)(C)[C@@H]1O)C)(=O)=O
Molecular Weight (g/mol) 411.44
Formula Weight 411.44
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.