missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HJ 805-2016 SVOC Internal Standard Mixture w/o 1,4-dichlorobenzene D4 4000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Acenaphthene D10∣Chrysene D12∣Naphthalene-d8∣Perylene D12∣Phenanthrene D10 |
| CAS | 15067-26-2, 1719-03-5, 1520-96-3, 1517-22-2 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Acenaphthene D10 4000 μg/mL∣Chrysene D12 4000 μg/mL∣Naphthalene-d8 4000 μg/mL∣Perylene D12 4000 μg/mL∣Phenanthrene D10 4000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?