Learn More
HJ 1153-2020 Aldehyde-DNPHs and Ketone-DNPHs Mixture 344 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30309127
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309127 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309127 Supplier Dr. Ehrenstorfer Supplier No. DREA50000349AL
 Restricted Item
Estimated Shipment: 07-12-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 958-60-1, 1019-57-4, 1567-89-1, 888-54-0, 1157-84-2, 1527-98-6
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile