missing translation for 'onlineSavingsMsg'
Learn More

ʟ-Histidine Monohydrochloride Monohydrate, Millipore™

Product Code. 10403021
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10403021 500 g 500g
10367040 25 g 25g
10280601 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10403021 Supplier Millipore Supplier No. 1043500500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5934-29-2

Specifications

Melting Point 254°C (dec.) (lit.)
MDL Number MFCD00151027
Synonym His, ʟ-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride, ʟ-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride
Density 1.49 g/cm3 at 20°C
Molecular Weight (g/mol) 209.63
Quantity 500 g
Grade Research
Chemical Name or Material ʟ-Histidine monohydrochloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.