missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HIN-1 Monoclonal Antibody (410219)
GREENER_CHOICE

Product Code. 15503842
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15503842 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15503842 Supplier Invitrogen™ Supplier No. MA524147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse HIN-1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

HIN-1, also known as High In Normal-1 or Secretoglobin Family 3A Member 1 (SCGB3A1), is a gene located on chromosome 5q33.3. It encodes a protein that is predominantly expressed in normal mammary epithelial cells and is thought to function in epithelial cell signaling, contributing to cellular differentiation and maintaining normal tissue architecture. The HIN-1 gene is often silenced or downregulated in various cancers, particularly breast cancer, due to hypermethylation of its promoter region, suggesting a potential tumor suppressor role. Loss of HIN-1 expression has been implicated in the progression of malignancies by disrupting normal cell signaling pathways, thereby facilitating oncogenesis. Given its significant downregulation in cancerous tissues, HIN-1 is being explored as a biomarker for early cancer detection and may provide insights into novel therapeutic strategies that aim to restore its expression and normal cellular functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIN-1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 410219
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SCGB3A1
Gene Accession No. Q96QR1
Gene Alias cytokine high in normal-1; cytokine HIN-1; High in normal 1; HIN1; HIN-1; LU105; pneumo secretory protein 2; PNSP2; PnSP-2; SCGB3A1; Secretoglobin family 3A member 1; secretoglobin, family 3A, member 1; UGRP2; UGRP2MGC87867; UNQ629/PRO1245; Uteroglobin-related protein 2
Gene Symbols SCGB3A1
Host Species Mouse
Immunogen E. coli-derived recombinant human HIN-1/SCGB3A1 Phe21-Gly104.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92304
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.