Learn More
Grant Instruments™ High-Speed Mini Centrifuge
Product Code. 13445056
Change view
Click to view available options
Volume (Metric):
12 x 1.5/2mL Microtubes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13445056 12 x 1.5/2mL Microtubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13445056 Supplier Grant Instruments™ Supplier No. MICROSPIN 12
  • ONLINE EXCLUSIVE PRICE
    1240.00 EUR
    1153.92 EUR / Pack of 1
    Save 86.08 EUR
    6% Off
View all promotions
Estimated Shipment: 27-11-2026
Microspin-12 compact high-speed benchtop microcentrifuge with a built-in rotor for 12 x 1.5/2ml microtubes. Ideal for biomedical laboratories.
  • Temperature setting range ambient +4ºC to +25ºC (Suitable to use in cold rooms)
  • Centrifugation speed: 100-14,500rpm, RCF 12,400 x g
  • Fixed angular aluminum rotor accommodates 12 x 1.5/2mL microtubes, supplied with a adapters for 0.2mL and 0.5mL tubes
  • Timed operation (1 to 30 minutes), with automatic switch-off
  • Very small footprint
TRUSTED_SUSTAINABILITY
Specifications
Temperature Range (Metric) Ambient +4° to 25°C
Volume (Metric) 12 x 1.5/2mL Microtubes
Holds 12 x 1.5/2 mL Microtubes
Speed 100 to 14500rpm
Timer 1 to 30 min.
Voltage 120 or 230 V