Learn More
DWK Life Sciences Wheaton™ Crimping Jaw Set for High Performance Crimping Tool
Product Code. 15778979 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
20 mm
For Use With (Application):
Crimping
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size For Use With (Application) Unit Size
15778979 20 mm Crimping Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15778979 Supplier DWK Life Sciences Supplier No. W225771
  • ONLINE EXCLUSIVE PRICE
    775.00 EUR
    693.70 EUR / Pack of 1
    Save 81.30 EUR
    10% Off
Estimated Shipment: 29-01-2027
Interchangeable jaws allow users to choose a jaw that best fits their needs.
  • Available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps
Specifications
Compatible Storage Tubes Tubes with 20 mm Caps
Operation Automated
Type Electronic Crimp Jaw
For Use With (Equipment) High performance crimping tool
Size 20 mm
For Use With (Application) Crimping