missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF1a Rabbit anti-Human, Clone: 2K1, Proteintech
Rabbit Recombinant Antibody
149.00 EUR - 439.00 EUR
Specifications
| Antigen | HIF1a |
|---|---|
| Clone | 2K1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17609816
|
Proteintech
80933-1-RR-20UL |
20 μL |
149.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17699806
|
Proteintech
80933-1-RR-100UL |
100 μL |
439.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIF1a antibody apecifically detetects HIF1a antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| HIF1a | |
| 1000 μg/mL | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 3091 | |
| HIF1a fusion protein Ag15198 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2K1 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Liquid | |
| Human | |
| BC012527 | |
| HIF1A | |
| IgG | |
| Protein A purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title