missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF1a Rabbit anti-Human, Clone: 2K1, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80933-1-RR-20UL
This item is not returnable.
View return policy
Description
HIF1a antibody apecifically detetects HIF1a antigen in Human Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| HIF1a | |
| 2K1 | |
| Unconjugated | |
| BC012527 | |
| Rabbit | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| HIF1A | |
| HIF1a fusion protein Ag15198 | |
| 20 μL | |
| 3091 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction