missing translation for 'onlineSavingsMsg'
Learn More

HIF-1 alpha Antibody - BSA Free, Novus Biologicals™

Product Code. 18725873 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
25µL
Quantity:
0.025 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Quantity
18725873 0.1mL -
18799973 25µL 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18725873 Supplier Novus Biologicals Supplier No. NB100134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 175 publications

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Chromatin Immunoprecipitation Sequencing, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY

Specifications

Dilution Western Blot 1:500 - 1:1000, Simple Western 1:100, ELISA, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:100 - 1:500, Immunoblotting, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 34277635), Dual RNAscope ISH-IHC
Gene Symbols HIF1A
Immunogen This HIF-1 alpha Antibody was developed against a fusion protein made to an internal sequence of human HIF-1 alpha (containing amino acids 432 - 528) [Uniprot# Q16665].
Regulatory Status RUO
Target Species Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.