missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Methanesulfonate, TRC

Product Code. 30012458
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012458 25 g 25g
30004036 5 g 5g
30007308 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012458 Supplier Toronto Research Chemicals Supplier No. TRCH29715525G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16156-50-6
Molecular Formula C7 H16 O3 S
Molecular Weight (g/mol) 180.265
Synonym Hexyl Mesylate, n-Hexyl Methanesulfonate, Methanesulfonic Acid Hexyl Ester
IUPAC Name hexyl methanesulfonate
SMILES CCCCCCOS(=O)(=O)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H16O3S/c1-3-4-5-6-7-10-11(2,8)9/h3-7H2,1-2H3
Formula Weight 180.082
Chemical Name or Material Hexyl Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.