Learn More
Hexane, Fraction from Petroleum, for GC Residue Analysis, Scharlab
Product Code. 11384174
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11384174 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11384174 Supplier Scharlab Supplier No. HE02232500
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Used in the preparation of specimens in the analysis of pesticides where solvent extraction is required. Hexane, Fraction from Petroleum, for GC Residue Analysis, Scharlab is an extraordinarily pure solvent, with controls that ensure the absence of peaks of over 3 ppt.
GC Residue Analysis Grade solvents are used to prepare specimens for pesticide analysis. The sample is evaporated, the residue is re-dissolved in the solvent and the solution is analyzed using gas chromatography. Evaporation concentrates both the residue and any impurities in the solvent itself, so this type of analysis requires extraordinarily pure solvents.
  • Controls performed by GC-ECD to ensure the absence of peaks of over 3 ppt (expressed as lindane), from low, medium and high boiling points (from 1,2,4-trichlorobenzene to decachlorobiphenyl), and also in the area around that of 2,4,5-trichlorobiphenyl.
  • These controls ensure that the quality of GC Residue Analysis solvents is suitable for organochlorinated, dioxin, furan and PCB pesticide residue analysis.
  • In pesticide residue analaysis, anhydrous sodium sulfate is normally used as a drying agent once the extraction process is performed, after separating the aqueous and organic phases.
Specifications
CAS 92112-69-1
Color Colorless
Appearance Clear
Density 0.67g/cm3
Quantity 2.5 L
Flash Point −22°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.38
Residue after Evaporation Max. 0.0001%
Molecular Formula C6H14
Vapor Pressure 160 hPa
MDL Number MFCD00009520
UN Number 1208
Water Max. 0.01%
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Boiling Point 65°C to 70°C
Solubility Information Insoluble in water
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
PubChem CID 8058
ChEBI CHEBI:29021
Formula Weight 86.18g/mol
Grade Fraction from petroleum
Chemical Name or Material Hexane