Learn More
Hexafluoroglutaric acid, 98%
Product Code. p-7023867
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463448 5 g 5g
11473448 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11463448 Supplier Thermo Scientific Alfa Aesar Supplier No. A17848.06
Add to basket
CAS: 376-73-8 | C5H2F6O4 | 240.06 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 376-73-8
Molecular Formula C5H2F6O4
Molecular Weight (g/mol) 240.06
MDL Number MFCD00004172
InChI Key CCUWGJDGLACFQT-UHFFFAOYSA-N
Synonym hexafluoroglutaric acid, perfluoroglutaric acid, hexafluoropentanedioic acid, pentanedioic acid, hexafluoro, glutaric acid, hexafluoro, pentanedioic acid, 2,2,3,3,4,4-hexafluoro, acmc-209iut, perfluoropentanedioic acid, glutaric acid, hexafluoro-8ci
PubChem CID 67827
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O
Specifications
Melting Point 92°C to 97°C
Boiling Point 134°C to 138°C (3 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 1801049
Sensitivity Hygroscopic
IUPAC Name hexafluoropentanedioic acid
Formula Weight 240.06
Percent Purity 98%
Chemical Name or Material Hexafluoroglutaric acid