missing translation for 'onlineSavingsMsg'
Learn More

Hexacosane, TRC

Product Code. 17997324
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17997324 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 17997324 Supplier Toronto Research Chemicals Supplier No. TRCH29050010G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 630-01-3
Molecular Formula C26 H54
Quantity 10 g
Synonym Hexacosane (6CI, 8CI, 9CI, ACI), n-Hexacosane, NSC 122457
InChI Formula InChI=1S/C26H54/c1-3-5-7-9-11-13-15-17-19-21-23-25-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-26H2,1-2H3
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC
IUPAC Name hexacosane
Molecular Weight (g/mol) 366.71
Formula Weight 366.4226
Chemical Name or Material n-Hexacosane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.