Learn More
Hexacarbonyltungsten, Sigma-Aldrich™
Product Code. 15814021
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814021 25 g 25g
15804021 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15814021 Supplier Sigma Aldrich Supplier No. 8008880025
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14040-11-0
Specifications
Melting Point 169°C to 171°C
Quantity 25 g
Density 2.65 g/cm3 at 20°C
Molecular Formula W(co)6
MDL Number MFCD00011462
Synonym Hexacarbonyltungsten, Tungsten carbonyl, Hexacarbonyltungsten
Molecular Weight (g/mol) 351.9
Grade Research
Chemical Name or Material Hexacarbonyltungsten