Learn More
Heptane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore
Product Code. 15845890
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15845890 25 g Glass 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15845890 Supplier Supelco Supplier No. 1183060025
 Restricted Item
Estimated Shipment: 11-11-2026
Ion pair reagents for analytical HPLC LiChropur
TRUSTED_SUSTAINABILITY
Specifications
CAS 22767-50-6
Density 391kg/m3
Packaging Glass
Molecular Formula C7H15NaO3S
Quantity 25 g
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;heptane-1-sulfonate
Molecular Weight (g/mol) 202.244
PubChem CID 23672332
Formula Weight 202.24
Percent Purity ≥99%
Grade Not Available
Physical Form Solid
Chemical Name or Material Heptane-1-sulfonic acid sodium salt