missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanoic Acid, MP Biomedicals

Product Code. 11451736 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451736 10 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11451736 Supplier MP Biomedicals Supplier No. 0520758305

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 506-12-7
Molecular Formula C17H34O2
Quantity 10 g
Synonym margaric acid, n-heptadecanoic acid, margarinic acid, n-heptadecylic acid, n-heptadecoic acid, heptadecylic acid, fatty acids, c16-18, unii-v987y9oz8l, normal-heptadecanoic acid, margarinate
InChI Key KEMQGTRYUADPNZ-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCC(=O)O
IUPAC Name heptadecanoic acid
Molecular Weight (g/mol) 270.457
PubChem CID 10465
ChEBI CHEBI:32365
Formula Weight 270.46
Chemical Name or Material Heptadecanoic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.