Learn More
Melford HEPES Buffered Saline Solution 2X
Product Code. 30018327
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018327 500 mL 500mL
30018326 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018327 Supplier Melford Supplier No. H17100500.0
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
2X Formulation:
NaCI: 280 mM
KCI: 10 mM
Na2HPO4: 1.5 mM
Dextrose: 12 mM
HEPES: 50 mM
pH: 7.0
Specifications
Chemical Name or Material HEPES Buffered Saline Solution 2 X
CAS 7647-14-5, 7447-40-7, 7558-79-4, 50-99-7, 7365-45-9, 7732-18-5
Quantity 500 mL