Learn More
Hirschmann™ Hematocrit Tubes, NH4-heparinized
Product Code. 30090385
Change view
Click to view available options
Capacity (Metric):
18 μL
Unit Size:
Pack of 5000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30090385 18 μL Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30090385 Supplier Hirschmann™ Supplier No. 9100418
  • ONLINE EXCLUSIVE PRICE
    447.00 EUR
    288.50 EUR / Pack of 5000
    Save 158.50 EUR
    35% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Excellent for centrifuging blood in the laboratory. Hirschmann™ Hematocrit Tubes are manufactured from disposable capillary glass.
  • Disposable
  • With ringmark at 60mm
  • NH4-heparinized
  • Color-code green
  • CE acc. IVD 98/79 EC
Specifications
Product Type Hematocrit Tube NH4-hep
Capacity (Metric) 18 μL
Diameter (Metric) Inner 0.5/0.6 mm
For Use With (Application) For centrifuging blood samples, Ammonium heparinized
Length (Metric) 75 mm
Certifications/Compliance CE, IVD 98/79 EC