Learn More
3M™ Helmet Back Plate Adaptor
Product Code. 15749857 Shop All 3M Products
Change view
Click to view available options
Style:
P3HV
Z3AF
Unit Size:
10 pairs
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Style Unit Size
15749857 Z3AF Each
15759957 P3HV 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15749857 Supplier 3M™ Supplier No. 7000103752
  • ONLINE EXCLUSIVE PRICE
    21.00 EUR
    12.66 EUR / Pack of 1
    Save 8.34 EUR
    39% Off
Estimated Shipment: 26-02-2027
3M™ Helmet Back Plate Adaptor is used to mount PELTOR Multi-Visors and PELTOR Ear Muffs fitted with the P3 mounts to 3M™ Safety Helmets.
  • Adaptor to connect 3M™ visors and multivisors to Safety Helmets for added face protection
  • Snaps easily into most safety helmets with universal accessory slots to help provide face protection
  • Adaptor for mounting visors or earmuffs
  • Snaps easily into the helmet's universal accessory socket to prevent these accessories from being damaged during use
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) Visors, Multivisors, and Protective Helmets
For Use With (Application) Connecting visors to helmets
Product Type Helmet Back Plate Adapter
Style Z3AF
Width (Metric) 25 mm