Learn More
Helios Italquartz Quartz Wool
Product Code. 11755694
Change view
Click to view available options
Quantity:
50 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11755694 50 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11755694 Supplier Helios Italquartz Supplier No. 39P00505
  • ONLINE EXCLUSIVE PRICE
    80.65 EUR
    72.54 EUR / Pack of 1
    Save 8.11 EUR
    10% Off
Estimated Shipment: 04-12-2026
For filtration and insulation in environments where high quartz purity properties are needed. Helios Italquartz™ Quartz Wool is chemically inert.
  • Supplied in 50g amounts
  • High temperature resistance
  • Resistant to corrosion
Specifications
Quantity 50 g
Material Quartz wool fiber