Learn More
Heidolph™ Hei-TORQUE Precision Series Overhead Stirrers
Product Code. 15301758 Shop All Heidolph Instruments Products
Change view
Click to view available options
Speed:
10 to 2000 rpm
10 to 400, 200 to 2000 rpm
Model:
Hei-TORQUE Precision 100
Hei-TORQUE Precision 200
Hei-TORQUE Precision 400
Torque:
100 Ncm
200 Ncm
400 Ncm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Model Torque Unit Size
15301758 10 to 2000 rpm Hei-TORQUE Precision 100 100 Ncm Each
15331758 10 to 400, 200 to 2000 rpm Hei-TORQUE Precision 400 400 Ncm Each
15311758 10 to 2000 rpm Hei-TORQUE Precision 200 200 Ncm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15301758 Supplier Heidolph™ Supplier No. 5016102000
  • ONLINE EXCLUSIVE PRICE
    2170.00 EUR
    1882.35 EUR / Pack of 1
    Save 287.65 EUR
    13% Off
View all promotions
Estimated Shipment: 04-12-2026
Heidolph™ Hei-TORQUE Precision Series Overhead Stirrers ideal for demanding tasks which have to be reproducible and documentable.
  • Accurate torque indication shows any viscosity changes
  • Constant speed even under changing loads
  • Newest motor generation provides maximum power at minimum noise level
  • Safe start and stop of operation via slide touch panel to avoid unintended stirring
  • RS 232 or USB interface to save all process data in digital file
  • Free software to automate and to save all process parameters
Digital 3.2 in. display for ease of operation:
  • Allows for pre-programmed profiles and saves profiles in memory
  • Interval operation
  • Watch rpm and torque graph life
  • Timer/Countdown/Real time setting
TRUSTED_SUSTAINABILITY
Specifications
Speed 10 to 2000 rpm
Display Digital
Voltage 230 V
Diameter (Metric) Shaft 10.5 mm
Description Dimensions: 241 x 93 x 328 mm
Model Hei-TORQUE Precision 100
No. of Speed Gears 1
Protection Class IP54
Torque 100 Ncm
Viscosity 60,000 mPas
Weight (Metric) 4.5 kg