Learn More
Koettermann™ Heavy Duty Table with Wheels
Product Code. 14825391
Change view
Click to view available options
Dimensions (L x W x H):
1200 x 600 x 750 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
14825391 1200 x 600 x 750 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14825391 Supplier Koettermann™ Supplier No. 50700014
  • ONLINE EXCLUSIVE PRICE
    2290.00 EUR
    1587.00 EUR / Pack of 1
    Save 703.00 EUR
    30% Off
Estimated Shipment: 29-01-2027
Add to basket
Add to basket
Koettermann™ Heavy Duty Table with Wheels includes a shelf for convenient material transport.
  • Capable of holding heavy loads
  • Includes shelf
Specifications
Product Type Mobile Table/Cart
Dimensions (L x W x H) 1200 x 600 x 750 mm
For Use With (Application) Carrying and transporting heavy loads
Includes Wheels, shelf