Learn More
S Murray and Co™ Heavy Duty Scissors
Product Code. 12952065
Change view
Click to view available options
Length (Metric):
180 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12952065 180 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12952065 Supplier S Murray and Co™ Supplier No. E916/05
  • ONLINE EXCLUSIVE PRICE
    5.90 EUR
    4.91 EUR / Pack of 1
    Save 0.99 EUR
    16% Off
Estimated Shipment: 21-12-2026
Use these general purpose scissors for cutting anything from bandages to stainless steel mesh. S Murray™ Heavy Duty Scissors are heavy duty but lightweight with stainless steel blades and robust construction.
  • Nickel plated steel
  • Straight with one sharp point
  • For cutting anything from bandages to stainless steel mesh
  • Autoclavable
Specifications
Product Type Heavy Duty Scissors
Blade Style Straight
Length (Metric) 180 mm
Material Stainless Steel