Learn More
Heavy Duty Folding Sack Truck
Product Code. 12955961
Encompass
Change view
Click to view available options
Load Bearing Capacity (Metric):
200 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity (Metric) unitSize
12955961 200 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12955961 Supplier Brand Not Defined Supplier No. FJ182H
A versatile sack truck which is lightweight yet strong, with fully welded tubular steel construction. Finished in a hard wearing epoxy powder coating. Easily folded for storage. Max load 200kg. Platform size 310mm wide x 330mm deep.
Specifications
Type Heavy-duty Folding Sack Truck
Load Bearing Capacity (Metric) 200 kg
Height (Metric) 960 mm
Weight (Metric) 10 kg
Width (Metric) 500 mm