Learn More
3M™ Heavy Duty EPDM Breathing Tube, SS-BT-55
Description
- Fixed-length breathing tube made from EPDM rubber
- Approved for use with 3M™ Powered Air Respirator PF-600E
- Suitable for use with specific 3M™ Promask™ Full Facepiece Reusable Respirator, FM3 and 3M™ Vision™ Full Facepiece Reusable Respirator with 3M™ Powered Air Respirator PF-600E
- Protective breathing tube covers available to lengthen service life of breathing tubes
Specifications
Specifications
| Material | EPDM Rubber |
| Product Type | Breathing Tube |
| For Use With (Equipment) | 3M™ PAPR Systems |
| Model | SS-BT-55 |