Learn More
Bibby Scientific™ Heating Mantle Spare Element
Product Code. 11708412
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11708412 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11708412 Supplier Bibby Scientific™ Supplier No. 0301215
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Capacity (Metric) 50 mL
For Use With EM Series 50mL
Voltage 230 V
Wattage 60w
Product Type Heating Mantle Spare Element
Frequency 50/60 Hz