Learn More
Electrothermal™ Heating Controller Temperature Probes
Product Code. 11544098 Shop All Thermo Scientific Products
Change view
Click to view available options
Temperature (Metric) Maximum:
400°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Maximum Unit Size
11544098 400°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11544098 Supplier Electrothermal™ Supplier No. 9996503
  • ONLINE EXCLUSIVE PRICE
    642.00 EUR
    422.48 EUR / Pack of 1
    Save 219.52 EUR
    34% Off
Estimated Shipment: 19-11-2026
Choose the appropriate temperature probe specific to the model of Electrothermal Digital Heating Controller (sold separately)
  • Temperature Probes
  • Suitable for use with Electrothermal Digital Heating Controllers
  • Controller sold separately
Specifications
Temperature (English) Maximum 752°F
Temperature (Metric) Maximum 400°C
Sensor Types PT100 Probe
Description Class A
Type Temperature Probe
For Use With (Application) MC810B Digital Controller