Learn More
Fisherbrand™ eCount™ Colony Counter
Product Code. 15253305 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Display Type:
Digital
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
15253305 Digital Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15253305
  • ONLINE EXCLUSIVE PRICE
    198.00 EUR
    151.47 EUR / Pack of 1
    Save 46.53 EUR
    23% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
Combines a multi-function electronic counter with a Sharpie™ pen for marking to prevent missing of double-counting colonies. The eCount™ Colony Counter has a light, ergonomic body that is balanced for easy manipulation. Different colored markers can be easily changed for marking versatility.
  • Sharpie pen for marking Petri dishes to prevent missing or double counting colonies
  • Save battery life
  • Verify each count with either beep or LED flash
  • First count up, then count down to verify accuracy
  • Display the total of all counts (up to 32 separate counts)
  • Keep up to 32 separate counts in memory
  • Disable counter so you can write totals and notes
  • Includes one fine-point Sharpie pen and weighted stand
Specifications
Product Type Colony Counter (Digital)
Display Type Digital
Length (English) 7.5 in.
Length (Metric) 19 cm