Learn More
Labconco™ Heated Product Shelves
Product Code. 10443285 Shop All Labconco Corporation Products
Change view
Click to view available options
Diameter (Metric):
254 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10443285 254 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10443285 Supplier Labconco™ Supplier No. 7509401
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Variable heat control shelves for freeze drying samples.
  • Three aluminum shelves with 6.35mm rims
  • Three stainless steel support rods
  • Nine clips for shelf height adjustment
  • Low voltage transformer with variable heat control to +40°C (+104°F)
  • Three rubber feet
Specifications
Type Heated Shelf
No. of Shelves 3
Temperature Range (Metric) Up to 40°C
Size Large
Diameter (English) 10 in.
Diameter (Metric) 254 mm
Height (English) 11.75 in.
Height (Metric) 298 mm
Voltage 230 V
Electrical Requirements 230 V 50/60 Hz
Frequency 50/60 Hz
Shipping Weight (English) 15.0 lb.
Shipping Weight (Metric) 6.8 kg