Learn More
3M™ Attest™ Biological Indicators
Product Code. 11973604
Change view
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11973604 100 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11973604 Supplier 3M™ Supplier No. 7000002535
  • ONLINE EXCLUSIVE PRICE
    1380.30 EUR
    387.84 EUR / Pack of 100
    Save 992.46 EUR
    71% Off
Estimated Shipment: 30-11-2026
Monitors the effectiveness of the steam sterilization process and provides results within 48 hours of incubation. 3M™ Attest™ Biological Indicators provide the best assurance of sterility by challenging the sterilizer with quantifiable, highly resistant spores.
  • Easy to use and interpret with visual color readout
  • Self-contained biological indicators significantly reduce possibility of contamination
  • Minimizes false positives
Specifications
Autoclavable Autoclavable
Certifications/Compliance EN886-1-2 et ISO 11138-1-3
Color Pink to Brown
Product Type Biological Indicator
Quantity 100 Pk.