Learn More
Buerkle™ Polypropylene Storage and Stacking Container
Product Code. 10298700 Shop All Bürkle Products
Change view
Click to view available options
Volume (Metric):
11 L
17 L
23 L
38 L
45 L
64 L
80 L
Dimensions (L x W x H):
400 x 300 x 120 mm
400 x 300 x 180 mm
400 x 300 x 220 mm
600 x 400 x 120 mm
600 x 400 x 180 mm
600 x 400 x 220 mm
600 x 400 x 320 mm
600 x 400 x 420 mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Dimensions (L x W x H) unitSize
10298700 11 L 400 x 300 x 120 mm Each
10657921 17 L 400 x 300 x 180 mm Each
11901471 23 L 400 x 300 x 220 mm Each
10507912 23 L 600 x 400 x 120 mm Each
10461231 38 L 600 x 400 x 180 mm Each
10701212 45 L 600 x 400 x 220 mm Each
10229180 64 L 600 x 400 x 320 mm Each
11921471 80 L 600 x 400 x 420 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10298700 Supplier Buerkle™ Supplier No. 34140111
  • ONLINE EXCLUSIVE PRICE
    28.95 EUR
    26.05 EUR / Pack of 1
    Save 2.90 EUR
    10% Off
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Buerkle™ Polypropylene Storage and Stacking Container with external reinforcement ribs.
  • Made of durable PP with external reinforcement ribs
  • Temperature resistant from -40°C to 90°C
  • According to Euro standards
  • Lid and transport rollers available as accessory
TRUSTED_SUSTAINABILITY
Specifications
Product Type Storage Case
Color Gray
Material Polypropylene
Certifications/Compliance According to Euro standards
Dimensions (D x W x H) Interior 367 x 267 x 116 mm
Dimensions (L x W x H) 400 x 300 x 120 mm
Height (Metric) 120 mm
Height (Metric) Interior 116 mm
Length (Metric) 400 mm
Length (Metric) Interior 367 mm
Volume (Metric) 11 L
Width (Metric) 300 mm
Width (Metric) Interior 267 mm