Learn More
Ratiolab™ HDPE Pasteur Pipet
Product Code. 10799823
Change view
Click to view available options
Volume (Metric):
1 mL
1.5 mL
3 mL
6 mL
10 mL
Length (Metric):
144 mm
148 mm
150 mm
155 mm
230 mm
300 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 400
Pack of 500
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) Sterility unitSize
10799823 1 mL 144 mm Non-sterile Pack of 500
10185742 1 mL 144 mm Sterile Pack of 500
10789823 1 mL 150 mm Sterile Pack of 500
10201351 1 mL 150 mm Non-sterile Pack of 500
10182772 1.5 mL 230 mm Non-sterile Pack of 400
10178111 3 mL 155 mm Non-sterile Pack of 500
12623165 6 mL 148 mm Non-sterile Pack of 400
10145792 10 mL 300 mm Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10799823 Supplier Ratiolab™ Supplier No. 2600181
Powering Proteomics
Use promo code 28003
View all promotions
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
Use these pipets for simple and safe liquid handling. Ratiolab™ HDPE Pasteur Pipets feature a low affinity surface which reduces the loss of cells and valuable proteins through binding.
  • No risk of cross-contamination or carry-over
  • No danger of glass breakage with the resulting danger of infectious material escaping
  • Transparent polyethylene construction
  • Inert to biological fluids and most acids
  • Soft, elastic bulb
  • Burr-free orifice for neat, clean and precise dispensing
  • Clearly visible embossed graduations
  • BSE-free
  • Can be used for food
Specifications
Material HDPE
Sterility Non-sterile
For Use With (Application) Food analysis
Includes Precisely Shaped Aspirator and Suction Ball
Quantity 500 Pack
Volume (Metric) 1 mL
Length (Metric) 144 mm
Type Pasteur Pipette