Learn More
Kartell™ Heavy-Walled Narrow Neck Carboy Bottles, HDPE
Description
Suitable for food stuffs; Autoclavable.
- Very resistant
- O-ring seal cap make it leakproof
- The cap can be wired down for added safety
- Includes carrying handles
- Suitable for foodstuff
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 25 L |
| Certifications/Compliance | FDA |
| Description | With O-ring seal cap |
| For Use With (Application) | Suitable for food stuffs |
| Includes | Closure, Carrying Handles |
| Material | HDPE |
| Mouth Style | Narrow |
| Neck Style | Narrow |
| No. per Pack | 4 |
| Show More |