Learn More
Kartell™ Male Bed Bottle with Cap
Product Code. 12051133
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12051133 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12051133 Supplier Kartell™ Supplier No. 0059100
  • ONLINE EXCLUSIVE PRICE
    4.00 EUR
    3.70 EUR / Pack of 1
    Save 0.30 EUR
    7% Off
Estimated Shipment: 27-11-2026
Designed with handle to allow it to be used in multiple positions. Kartell™ Male Bed Bottle with Cap ensures a complete and secure seal.
  • For hospital use
  • CE Marked and IVD Certified
  • Includes cap
  • Constructed of polyethylene
Specifications
Product Type Male Bed Bottle with Cap
Color White
Material Polyethylene
For Use With (Application) Hospital use
Volume (Metric) 1000 mL