Learn More
Karl Hecht™ HDPE Funnels
Product Code. 10688871
Change view
Click to view available options
Diameter (Metric)Top:
12 cm
20 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Unit Size
10688871 12 cm Each
10769842 20 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10688871 Supplier Karl Hecht™ Supplier No. 49050012
  • ONLINE EXCLUSIVE PRICE
    4.60 EUR
    1.98 EUR / Pack of 1
    Save 2.62 EUR
    56% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Manufactured from HDPE for optimum strength and chemical resistance.
  • White in color
  • For general laboratory use
  • Durable polyethylene
  • Available in different diameters and stem lengths
Specifications
Diameter (Metric)Top 12 cm
Material HDPE
Color White
Length (Metric) Stem 60 mm
Product Type Funnel