Learn More
Brand™ HDPE Atomizers
Product Code. 12015302 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
850 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12015302 850 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12015302 Supplier Brand™ Supplier No. 144190
  • ONLINE EXCLUSIVE PRICE
    63.00 EUR
    59.75 EUR / Pack of 5
    Save 3.25 EUR
    5% Off
Estimated Shipment: 14-12-2026
By turning the nozzle, delivery can be varied from a fine spray to a solid stream.
  • Opaque white in color with red accents
  • Provided in packs of 5
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 850 mL
Color White
Material HDPE
No. per Pack 5/Pk.
Product Type Atomizer