Learn More
HBTU, TRC
Product Code. 30390346
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390346 1 g
30402878 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30390346 Supplier Toronto Research Chemicals Supplier No. TRCH1075001G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 94790-37-1
Molecular Formula C11H16F6N5OP
Molecular Weight (g/mol) 379.24
Synonym 1-[Bis(dimethylamino)methylene]-1H-benzotriazolium Hexafluorophosphate(1-) 3-Oxide, N-HBTU
IUPAC Name [benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)On1nnc2ccccc12.F[P-](F)(F)(F)(F)F
Specifications
Quantity 1 g
InChI Formula InChI=1S/C11H16N5O.F6P/c1-14(2)11(15(3)4)17-16-10-8-6-5-7-9(10)12-13-16;1-7(2,3,4,5)6/h5-8H,1-4H3;/q+1;-1
Formula Weight 379.0997
Chemical Name or Material HBTU